Assay Details
Gene Symbol: Prkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008926.4 | NP_032952.3 | 8 - 9 | 1259 | 93 | |
GenBank mRNA | AK038739.1 | - | 8 - 9 | 1243 | 93 | |
AK081422.1 | - | 8 - 9 | 1260 | 93 | ||
BC113205.1 | - | 8 - 9 | 1170 | 40047751 | 93 | |
L12460.1 | - | 8 - 9 | 1531 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
