Assay Details
Gene Symbol: Mapkapk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010765.2 | NP_034895.1 | 7 - 8 | 1269 | 68 | |
GenBank mRNA | AF039840.1 | - | 7 - 8 | 1283 | 68 | |
AY533679.1 | - | 7 - 8 | 577 | 68 | ||
AY533680.2 | - | 7 - 8 | 577 | 68 | ||
AY533681.1 | - | 2 - 3 | 130 | 68 | ||
AY533682.1 | - | 2 - 3 | 130 | 68 | ||
BC019184.1 | - | 7 - 8 | 691 | 5010305 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
