Assay Details
Gene Symbol: Atp6v0a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011596.5 | NP_035726.2 | 1640 | 71 | ||
GenBank mRNA | AB022323.1 | - | 1606 | 71 | ||
AF218252.1 | - | 1625 | 71 | |||
AK155055.1 | - | 1617 | 71 | |||
AK170122.1 | - | 2137 | 71 | |||
BC013056.1 | - | 3 - 4 | 372 | 71 | ||
BC018369.1 | - | 1593 | 71 | |||
BC108991.1 | - | 1537 | 40047832 | 71 | ||
BC108992.1 | - | 1537 | 40047833 | 71 | ||
BC112905.1 | - | 1600 | 3670722 | 71 | ||
M31226.1 | - | 1536 | 71 | |||
X55184.1 | - | 1537 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
