Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 17 - 18 | 1796 | 109 | |
NM_007982.2 | NP_032008.2 | 16 - 17 | 1641 | 109 | ||
GenBank mRNA | AB011499.1 | - | 17 - 18 | 1452 | 109 | |
AB030035.1 | - | 16 - 17 | 1368 | 109 | ||
AK079821.1 | - | 17 - 18 | 1893 | 109 | ||
AK220543.1 | - | 16 - 17 | 1641 | 109 | ||
BC030180.1 | - | 17 - 18 | 1726 | 109 | ||
JN971016.1 | - | 16 - 17 | 1368 | 109 | ||
M95408.1 | - | 15 - 16 | 1438 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
