Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 1 - 2 | 184 | 135 | |
NM_007982.2 | NP_032008.2 | 1 - 2 | 184 | 135 | ||
GenBank mRNA | AK005468.1 | - | 1 - 2 | 208 | 135 | |
AK079821.1 | - | 1 - 2 | 123 | 135 | ||
AK135490.1 | - | 1 - 2 | 120 | 135 | ||
AK220543.1 | - | 1 - 2 | 184 | 135 | ||
BC030180.1 | - | 1 - 2 | 114 | 135 | ||
CJ050166.1 | - | 1 - 2 | 142 | 135 | ||
DQ478404.1 | - | 1 - 2 | 71 | 135 | ||
DQ478405.1 | - | 1 - 2 | 71 | 135 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK005468.1, AK079821.1, DQ478404.1, DQ478405.1, NM_001130409.1, NM_007982.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
