Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 21 - 22 | 2203 | 152 | |
NM_007982.2 | NP_032008.2 | 20 - 21 | 2048 | 152 | ||
GenBank mRNA | AB011499.1 | - | 21 - 22 | 1859 | 152 | |
AB030035.1 | - | 19 - 20 | 1674 | 152 | ||
AK079821.1 | - | 21 - 22 | 2300 | 152 | ||
AK220543.1 | - | 20 - 21 | 2048 | 152 | ||
BC030180.1 | - | 21 - 22 | 2133 | 152 | ||
JN971016.1 | - | 20 - 21 | 1775 | 152 | ||
M95408.1 | - | 19 - 20 | 1845 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
