Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 28 - 29 | 3027 | 64 | |
NM_007982.2 | NP_032008.2 | 27 - 28 | 2872 | 64 | ||
GenBank mRNA | AB011499.1 | - | 28 - 29 | 2683 | 64 | |
AB030035.1 | - | 25 - 26 | 2293 | 64 | ||
AK079821.1 | - | 28 - 29 | 3124 | 64 | ||
AK220543.1 | - | 27 - 28 | 2872 | 64 | ||
BC030180.1 | - | 28 - 29 | 2957 | 64 | ||
JN971016.1 | - | 27 - 28 | 2599 | 64 | ||
M95408.1 | - | 26 - 27 | 2669 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
