Assay Details
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130409.1 | NP_001123881.1 | 5 - 6 | 825 | 94 | |
NM_007982.2 | NP_032008.2 | 4 - 5 | 670 | 94 | ||
GenBank mRNA | AB011499.1 | - | 2 - 3 | 358 | 94 | |
AB030035.1 | - | 2 - 3 | 358 | 94 | ||
AK079821.1 | - | 5 - 6 | 922 | 94 | ||
AK220543.1 | - | 4 - 5 | 670 | 94 | ||
BC030180.1 | - | 5 - 6 | 755 | 94 | ||
JN971016.1 | - | 2 - 3 | 358 | 94 | ||
M95408.1 | - | 3 - 4 | 467 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
