Assay Details
Gene Symbol: Mtus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005863.2 | NP_001005863.1 | 1 - 2 | 249 | 97 | |
GenBank mRNA | AK040710.1 | - | 1 - 2 | 242 | 97 | |
AK143781.1 | - | 1 - 2 | 250 | 97 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK040710.1, AK143781.1, NM_001005863.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
