Assay Details
Gene Symbol: Oas1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033541.4 | NP_291019.1 | 1 - 2 | 86 | 102 | |
GenBank mRNA | AK028986.1 | - | 1 - 2 | 87 | 102 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK028986.1, NM_033541.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
