Assay Details
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029749.2 | NP_084025.2 | 12 - 13 | 1503 | 77 | |
GenBank mRNA | AY336567.1 | - | 11 - 12 | 1376 | 77 | |
BC137852.1 | - | 11 - 12 | 1396 | 8860871 | 77 | |
BM940384.1 | - | 4 - 5 | 418 | 77 | ||
CF532651.1 | - | 3 - 4 | 402 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
