Assay Details
Gene Symbol: Usp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033202.3 | NP_001028374.1 | 4 - 5 | 575 | 81 | |
GenBank mRNA | AK144578.1 | - | 4 - 5 | 576 | 81 | |
BC038606.1 | - | 4 - 5 | 597 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
