Assay Details
Gene Symbol: Hnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009327.3 | NP_033353.2 | 2 - 3 | 749 | 80 | |
GenBank mRNA | AK141225.1 | - | 2 - 3 | 556 | 80 | |
AK144051.1 | - | 2 - 3 | 756 | 80 | ||
M57966.1 | - | 729 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
