Assay Details
Gene Symbol: Hnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009327.3 | NP_033353.2 | 4 - 5 | 1176 | 63 | |
GenBank mRNA | AK141225.1 | - | 4 - 5 | 982 | 63 | |
AK144051.1 | - | 4 - 5 | 1184 | 63 | ||
BC080698.1 | - | 4 - 5 | 1178 | 30471380 | 63 | |
M57966.1 | - | 1156 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
