Assay Details
Gene Symbol: Hnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009327.3 | NP_033353.2 | 6 - 7 | 1529 | 155 | |
GenBank mRNA | AK141225.1 | - | 6 - 7 | 1335 | 155 | |
AK144051.1 | - | 6 - 7 | 1537 | 155 | ||
BC080698.1 | - | 6 - 7 | 1531 | 30471380 | 155 | |
M57966.1 | - | 1509 | 155 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
