Assay Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080706.1 | NP_001074175.1 | 21 - 22 | 3345 | 102 | |
GenBank mRNA | AB287295.1 | - | 21 - 22 | 3070 | 102 | |
AK141888.1 | - | 15 - 16 | 3792 | 102 | ||
BC094345.1 | - | 2 - 3 | 462 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
