Assay Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080706.1 | NP_001074175.1 | 30 - 31 | 4614 | 138 | |
GenBank mRNA | AB287295.1 | - | 30 - 31 | 4339 | 138 | |
AK088840.1 | - | 5 - 6 | 543 | 138 | ||
BC094345.1 | - | 11 - 12 | 1731 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
