Assay Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080706.1 | NP_001074175.1 | 36 - 37 | 5547 | 94 | |
GenBank mRNA | AB287295.1 | - | 36 - 37 | 5272 | 94 | |
AK088840.1 | - | 11 - 12 | 1476 | 94 | ||
BC019619.1 | - | 1 - 2 | 138 | 94 | ||
BC038277.1 | - | 3 - 4 | 543 | 94 | ||
BC094345.1 | - | 17 - 18 | 2664 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
