Assay Details
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080706.1 | NP_001074175.1 | 9 - 10 | 1301 | 110 | |
GenBank mRNA | AB287295.1 | - | 9 - 10 | 1026 | 110 | |
AK141888.1 | - | 3 - 4 | 1748 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
