Assay Details
Gene Symbol: Usp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033202.3 | NP_001028374.1 | 8 - 9 | 884 | 72 | |
GenBank mRNA | AK078164.1 | - | 4 - 5 | 236 | 72 | |
AK144578.1 | - | 8 - 9 | 885 | 72 | ||
AK162269.1 | - | 1 - 2 | 54 | 72 | ||
BC038606.1 | - | 8 - 9 | 906 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
