Assay Details
Gene Symbol: 2300005B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081961.1 | NP_001075430.1 | 2 - 3 | 172 | 60 | |
GenBank mRNA | AB272582.1 | - | 2 - 3 | 160 | 60 | |
AK009031.1 | - | 1 - 2 | 95 | 60 | ||
BC115610.1 | - | 2 - 3 | 172 | 40102714 | 60 | |
BC115611.1 | - | 2 - 3 | 172 | 40102716 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165234 | BC115610.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165234 | BC115611.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165234 | NM_001081961.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2165234 | AK009031.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702606 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213177 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886326 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165234 | BC115610.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165234 | BC115611.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165234 | NM_001081961.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2165234 | AK009031.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339364 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339374 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339359 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339373 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12136209 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339364 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339344 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339352 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339369 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339342 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12199980 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12136198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339362 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12136203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339347 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339356 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339339 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339358 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339377 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339350 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12199973 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339384 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339357 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339382 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30339372 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10429513 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429514 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429511 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10429512 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10429510 | - | - |
Affymetrix Mouse Clariom S Array | PSR1500012961.mm.1 | TC1500001716.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012961.mm.1 | NM_001081961.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012961.mm.1 | NM_001081961.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 382236 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3840047 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2700364 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2700364 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2700364 | - | - |
