Assay Details
Gene Symbol: Ap2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077264.1 | NP_001070732.1 | 17 - 18 | 2401 | 79 | |
NM_007458.2 | NP_031484.1 | 18 - 19 | 2467 | 79 | ||
GenBank mRNA | AK135424.1 | - | 17 - 18 | 2330 | 79 | |
AK155515.1 | - | 17 - 18 | 2338 | 79 | ||
BC031433.1 | - | 18 - 19 | 2417 | 4481643 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
