Assay Details
Gene Symbol: Cpsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164173.1 | NP_001157645.1 | 5 - 6 | 654 | 73 | |
NM_053193.2 | NP_444423.1 | 5 - 6 | 654 | 73 | ||
GenBank mRNA | AF322193.1 | - | 5 - 6 | 625 | 73 | |
AK155648.1 | - | 5 - 6 | 595 | 73 | ||
AK155806.1 | - | 5 - 6 | 576 | 73 | ||
BC011458.1 | - | 1 - 2 | 93 | 73 | ||
BC049941.1 | - | 5 - 6 | 476 | 73 | ||
BC056388.1 | - | 5 - 6 | 654 | 6854851 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
