Assay Details
Gene Symbol: Eif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133916.2 | NP_598677.1 | 9 - 10 | 1421 | 68 | |
GenBank mRNA | AK089978.1 | - | 9 - 10 | 1409 | 68 | |
AK167618.1 | - | 9 - 10 | 1408 | 68 | ||
AK170938.1 | - | 9 - 10 | 1421 | 68 | ||
AK211352.1 | - | 3 - 4 | 230 | 68 | ||
AK219469.1 | - | 3 - 4 | 232 | 68 | ||
BC007175.1 | - | 8 - 9 | 880 | 68 | ||
BC023767.1 | - | 9 - 10 | 1392 | 5343631 | 68 | |
BC031704.1 | - | 9 - 10 | 1398 | 4023277 | 68 | |
BC051065.1 | - | 9 - 10 | 1381 | 5343631 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
