Assay Details
Gene Symbol: Gigyf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031408.2 | NP_113596.2 | 15 - 16 | 1929 | 56 | |
GenBank mRNA | AK204736.1 | - | 1 - 2 | 38 | 56 | |
AK220289.1 | - | 8 - 9 | 938 | 56 | ||
AY176042.1 | - | 15 - 16 | 1945 | 56 | ||
BC052875.1 | - | 9 - 10 | 1033 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
