Assay Details
Gene Symbol: Jak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048177.2 | NP_001041642.1 | 12 - 13 | 2121 | 154 | |
NM_008413.3 | NP_032439.2 | 13 - 14 | 2204 | 154 | ||
GenBank mRNA | AK087255.1 | - | 12 - 13 | 2088 | 154 | |
BC054807.1 | - | 12 - 13 | 1884 | 6838318 | 154 | |
BC059834.1 | - | 12 - 13 | 1884 | 6838318 | 154 | |
L16956.1 | - | 12 - 13 | 1873 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
