Assay Details
Gene Symbol: Jak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048177.2 | NP_001041642.1 | 16 - 17 | 2618 | 80 | |
NM_008413.3 | NP_032439.2 | 17 - 18 | 2701 | 80 | ||
GenBank mRNA | BC054807.1 | - | 16 - 17 | 2381 | 6838318 | 80 |
BC059834.1 | - | 16 - 17 | 2381 | 6838318 | 80 | |
L16956.1 | - | 16 - 17 | 2370 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
