Assay Details
Gene Symbol: Slc30a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178651.3 | NP_848766.2 | 3 - 4 | 472 | 117 | |
GenBank mRNA | AK028885.1 | - | 3 - 4 | 439 | 117 | |
AK035541.1 | - | 3 - 4 | 459 | 117 | ||
AK082853.1 | - | 3 - 4 | 474 | 117 | ||
AK143277.1 | - | 3 - 4 | 473 | 117 | ||
AY682914.1 | - | 4 - 5 | 465 | 117 | ||
BC027806.1 | - | 3 - 4 | 234 | 117 | ||
BC078440.1 | - | 1 - 2 | 49 | 117 | ||
BC141008.1 | - | 4 - 5 | 438 | 9055620 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
