Assay Details
Gene Symbol: Kcnab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289450.1 | NP_001276379.1 | 7 - 8 | 468 | 100 | |
NM_010597.4 | NP_034727.3 | 6 - 7 | 884 | 100 | ||
GenBank mRNA | AF033003.1 | - | 6 - 7 | 1239 | 100 | |
AK004666.1 | - | 7 - 8 | 452 | 100 | ||
AK015412.1 | - | 8 - 9 | 724 | 100 | ||
AK031362.1 | - | 6 - 7 | 885 | 100 | ||
AK138467.1 | - | 6 - 7 | 873 | 100 | ||
AK153200.1 | - | 7 - 8 | 470 | 100 | ||
BC014701.1 | - | 6 - 7 | 609 | 4482268 | 100 | |
U65591.1 | - | 6 - 7 | 480 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
