Assay Details
Gene Symbol: Ift80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026641.2 | NP_080917.1 | 10 - 11 | 1253 | 155 | |
GenBank mRNA | AK042231.1 | - | 10 - 11 | 1178 | 155 | |
AK129339.1 | - | 10 - 11 | 1181 | 155 | ||
AK147305.1 | - | 9 - 10 | 4780 | 155 | ||
BC013814.1 | - | 5 - 6 | 542 | 155 | ||
BC034101.1 | - | 10 - 11 | 1148 | 4501451 | 155 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
