Assay Details
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013752.3 | NP_038780.3 | 10 - 11 | 1507 | 83 | |
GenBank mRNA | AF076687.1 | - | 10 - 11 | 1501 | 83 | |
AF092840.1 | - | 10 - 11 | 1490 | 83 | ||
AK031933.1 | - | 10 - 11 | 1493 | 83 | ||
AK132302.1 | - | 10 - 11 | 1549 | 84 | ||
AK134805.1 | - | 10 - 11 | 1543 | 83 | ||
AK134960.1 | - | 10 - 11 | 1535 | 83 | ||
AK159790.1 | - | 10 - 11 | 1514 | 83 | ||
AK160614.1 | - | 10 - 11 | 1474 | 83 | ||
BC003719.1 | - | 10 - 11 | 1516 | 83 | ||
BC013190.1 | - | 10 - 11 | 1507 | 83 | ||
BC044773.2 | - | 10 - 11 | 1506 | 5067851 | 83 | |
BC055061.1 | - | 10 - 11 | 1499 | 6528890 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
