Assay Details
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013752.3 | NP_038780.3 | 7 - 8 | 996 | 129 | |
GenBank mRNA | AB016988.1 | - | 7 - 8 | 918 | 129 | |
AF076687.1 | - | 7 - 8 | 990 | 129 | ||
AF092840.1 | - | 7 - 8 | 979 | 129 | ||
AK031933.1 | - | 7 - 8 | 982 | 129 | ||
AK132302.1 | - | 7 - 8 | 1037 | 129 | ||
AK134805.1 | - | 7 - 8 | 1032 | 129 | ||
AK134960.1 | - | 7 - 8 | 1024 | 129 | ||
AK159790.1 | - | 7 - 8 | 1003 | 129 | ||
AK160614.1 | - | 7 - 8 | 963 | 129 | ||
BC003719.1 | - | 7 - 8 | 1005 | 129 | ||
BC013190.1 | - | 7 - 8 | 996 | 129 | ||
BC044773.2 | - | 7 - 8 | 995 | 5067851 | 129 | |
BC055061.1 | - | 7 - 8 | 988 | 6528890 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
