Assay Details
Gene Symbol: Kit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122733.1 | NP_001116205.1 | 10 - 11 | 1719 | 195 | |
NM_021099.3 | NP_066922.2 | 10 - 11 | 1703 | 183 | ||
GenBank mRNA | AK046795.1 | - | 10 - 11 | 1711 | 195 | |
AK047010.1 | - | 10 - 11 | 1715 | 195 | ||
AK145462.1 | - | 7 - 8 | 944 | 183 | ||
AY536430.1 | - | 10 - 11 | 1666 | 183 | ||
AY536431.1 | - | 10 - 11 | 1666 | 183 | ||
BC026713.1 | - | 10 - 11 | 1698 | 195 | ||
BC075716.1 | - | 10 - 11 | 1704 | 3673641 | 195 | |
Y00864.1 | - | 10 - 11 | 1670 | 183 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
