Assay Details
Gene Symbol: Ociad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026950.3 | NP_081226.1 | 2 - 3 | 232 | 94 | |
GenBank mRNA | AK007617.1 | - | 2 - 3 | 186 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
