Assay Details
Gene Symbol: Ociad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026950.3 | NP_081226.1 | 5 - 6 | 432 | 90 | |
GenBank mRNA | AK007617.1 | - | 5 - 6 | 386 | 90 | |
BC044883.1 | - | 5 - 6 | 351 | 4971755 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
