Assay Details
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025951.3 | NP_080227.2 | 1 - 2 | 396 | 115 | |
GenBank mRNA | AF411321.1 | - | 1 - 2 | 251 | 115 | |
AK011751.1 | - | 1 - 2 | 359 | 115 | ||
AK035523.1 | - | 1 - 2 | 381 | 115 | ||
AK170730.1 | - | 1 - 2 | 380 | 115 | ||
BC062144.1 | - | 1 - 2 | 303 | 6335661 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
