Assay Details
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025951.3 | NP_080227.2 | 8 - 9 | 1327 | 157 | |
NM_028744.3 | NP_083020.2 | 8 - 9 | 1258 | 157 | ||
GenBank mRNA | AF411321.1 | - | 8 - 9 | 1182 | 157 | |
AK011751.1 | - | 8 - 9 | 1290 | 157 | ||
AK016754.1 | - | 8 - 9 | 1221 | 157 | ||
AK035523.1 | - | 8 - 9 | 1312 | 157 | ||
AK170730.1 | - | 8 - 9 | 1311 | 157 | ||
AY148879.1 | - | 1221 | 157 | |||
BC010257.1 | - | 335 | 157 | |||
BC062144.1 | - | 8 - 9 | 1234 | 6335661 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
