Assay Details
Gene Symbol: Acox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030721.2 | NP_109646.2 | 16 - 17 | 2019 | 63 | |
GenBank mRNA | AK028818.1 | - | 16 - 17 | 2440 | 63 | |
AK053960.1 | - | 16 - 17 | 2016 | 63 | ||
AK128922.1 | - | 16 - 17 | 1979 | 63 | ||
AK154112.1 | - | 15 - 16 | 2110 | 63 | ||
AK171598.1 | - | 2002 | 63 | |||
AK211732.1 | - | 1 - 2 | 66 | 63 | ||
BC044725.2 | - | 16 - 17 | 2039 | 5374281 | 63 | |
BC055019.1 | - | 16 - 17 | 2019 | 4162223 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
