Assay Details
Gene Symbol: Dcp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133761.3 | NP_598522.3 | 2 - 3 | 298 | 96 | |
GenBank mRNA | AJ344447.1 | - | 2 - 3 | 298 | 96 | |
AK047460.1 | - | 2 - 3 | 229 | 96 | ||
AK171707.1 | - | 2 - 3 | 222 | 96 | ||
BC066173.1 | - | 2 - 3 | 251 | 30049516 | 96 | |
DV651307.1 | - | 2 - 3 | 328 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
