Assay Details
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008069.4 | NP_032095.1 | 1 - 2 | 82 | 70 | |
GenBank mRNA | BC130258.1 | - | 1 - 2 | 86 | 40142758 | 70 |
BC157920.1 | - | 1 - 2 | 148 | 9007350 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
