Assay Details
Gene Symbol: Cwh43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181323.2 | NP_851840.1 | 5 - 6 | 1002 | 69 | |
GenBank mRNA | AK035550.1 | - | 5 - 6 | 853 | 69 | |
AK081976.1 | - | 5 - 6 | 1002 | 69 | ||
BC016523.1 | - | 5 - 6 | 830 | 3584006 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
