Assay Details
Gene Symbol: Camsap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163749.1 | NP_001157221.1 | 5 - 6 | 1122 | 89 | |
NM_027171.3 | NP_081447.2 | 5 - 6 | 1122 | 89 | ||
GenBank mRNA | AK081728.1 | - | 5 - 6 | 897 | 89 | |
AK220401.1 | - | 1 - 2 | 267 | 89 | ||
BC043081.1 | - | 5 - 6 | 778 | 89 | ||
BC048787.1 | - | 5 - 6 | 770 | 6489361 | 89 | |
KC426930.1 | - | 7 - 8 | 841 | 89 | ||
KC426931.1 | - | 7 - 8 | 841 | 89 | ||
KC426932.1 | - | 6 - 7 | 808 | 89 | ||
KC426933.1 | - | 6 - 7 | 808 | 89 | ||
KC426934.1 | - | 6 - 7 | 793 | 89 | ||
KC426935.1 | - | 6 - 7 | 793 | 89 | ||
KC426936.1 | - | 5 - 6 | 760 | 89 | ||
KC426937.1 | - | 5 - 6 | 760 | 89 | ||
KC426938.1 | - | 7 - 8 | 841 | 89 | ||
KC426939.1 | - | 7 - 8 | 841 | 89 | ||
KC426940.1 | - | 6 - 7 | 808 | 89 | ||
KC426941.1 | - | 6 - 7 | 808 | 89 | ||
KC426942.1 | - | 6 - 7 | 793 | 89 | ||
KC426943.1 | - | 6 - 7 | 793 | 89 | ||
KC426944.1 | - | 5 - 6 | 760 | 89 | ||
KC426945.1 | - | 5 - 6 | 760 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
