Assay Details
Gene Symbol: Gckr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144909.1 | NP_659158.1 | 12 - 13 | 1108 | 90 | |
GenBank mRNA | AK149377.1 | - | 10 - 11 | 3007 | 90 | |
BC012412.1 | - | 12 - 13 | 1108 | 4159892 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
