Assay Details
Gene Symbol: Bag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026121.3 | NP_080397.1 | 3 - 4 | 670 | 78 | |
GenBank mRNA | AF332863.1 | - | 3 - 4 | 663 | 78 | |
AK010765.1 | - | 3 - 4 | 661 | 78 | ||
AK136899.1 | - | 3 - 4 | 660 | 78 | ||
AK160316.1 | - | 3 - 4 | 662 | 78 | ||
AK162658.1 | - | 3 - 4 | 657 | 78 | ||
BC037239.1 | - | 3 - 4 | 668 | 78 | ||
BC058518.1 | - | 3 - 4 | 653 | 5717401 | 78 | |
BC138201.1 | - | 3 - 4 | 748 | 8861220 | 78 | |
BC145986.1 | - | 3 - 4 | 748 | 40131289 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
