Assay Details
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK156746.1 | - | 3 - 4 | 1218 | 93 | |
AK170040.1 | - | 3 - 4 | 1281 | 93 | ||
AK172238.1 | - | 3 - 4 | 1244 | 93 | ||
BC036336.1 | - | 2 - 3 | 810 | 93 | ||
BC064447.1 | - | 3 - 4 | 1256 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
