Assay Details
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK079952.1 | - | 1 - 2 | 136 | 81 | |
AK156746.1 | - | 4 - 5 | 1375 | 81 | ||
AK170040.1 | - | 4 - 5 | 1438 | 81 | ||
AK172238.1 | - | 4 - 5 | 1401 | 81 | ||
BC036336.1 | - | 3 - 4 | 967 | 81 | ||
BC064447.1 | - | 4 - 5 | 1413 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
