Assay Details
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK079952.1 | - | 3 - 4 | 801 | 131 | |
AK156746.1 | - | 6 - 7 | 2040 | 131 | ||
AK170040.1 | - | 6 - 7 | 2103 | 131 | ||
BC036336.1 | - | 5 - 6 | 1632 | 131 | ||
BC064447.1 | - | 6 - 7 | 2078 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
