Assay Details
Gene Symbol: Slc26a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145947.2 | NP_666059.2 | 12 - 13 | 1552 | 100 | |
GenBank mRNA | AF345194.1 | - | 11 - 12 | 1454 | 100 | |
AK054327.1 | - | 12 - 13 | 1553 | 100 | ||
BC026928.1 | - | 12 - 13 | 1531 | 4020760 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
