Assay Details
Gene Symbol: Gria2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039195.1 | NP_001034284.1 | 10 - 11 | 1887 | 102 | |
NM_001083806.1 | NP_001077275.1 | 10 - 11 | 1887 | 102 | ||
NM_013540.2 | NP_038568.2 | 10 - 11 | 1887 | 102 | ||
GenBank mRNA | AB109210.1 | - | 10 - 11 | 1480 | 102 | |
AB109211.1 | - | 10 - 11 | 1480 | 102 | ||
AB111957.1 | - | 10 - 11 | 1480 | 102 | ||
AK014389.1 | - | 10 - 11 | 1932 | 102 | ||
AK044574.1 | - | 10 - 11 | 1819 | 102 | ||
AK046861.1 | - | 10 - 11 | 1887 | 102 | ||
AK086614.1 | - | 10 - 11 | 1821 | 102 | ||
AK134572.1 | - | 10 - 11 | 1817 | 102 | ||
AK134647.1 | - | 9 - 10 | 1333 | 101 | ||
AK134772.1 | - | 9 - 10 | 1458 | 102 | ||
AK141525.1 | - | 10 - 11 | 1863 | 102 | ||
BC066193.1 | - | 10 - 11 | 1675 | 102 | ||
X57498.1 | - | 10 - 11 | 1682 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
