Assay Details
Gene Symbol: Gria2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039195.1 | NP_001034284.1 | 5 - 6 | 1122 | 89 | |
NM_001083806.1 | NP_001077275.1 | 5 - 6 | 1122 | 89 | ||
NM_013540.2 | NP_038568.2 | 5 - 6 | 1122 | 89 | ||
GenBank mRNA | AB109210.1 | - | 5 - 6 | 715 | 89 | |
AB109211.1 | - | 5 - 6 | 715 | 89 | ||
AK014389.1 | - | 5 - 6 | 1167 | 89 | ||
AK043490.1 | - | 5 - 6 | 1095 | 89 | ||
AK046861.1 | - | 5 - 6 | 1122 | 89 | ||
AK086614.1 | - | 5 - 6 | 1056 | 89 | ||
AK134572.1 | - | 5 - 6 | 1053 | 89 | ||
AK134647.1 | - | 4 - 5 | 568 | 89 | ||
AK134772.1 | - | 4 - 5 | 693 | 89 | ||
AK141525.1 | - | 5 - 6 | 1097 | 89 | ||
AK163941.1 | - | 5 - 6 | 1115 | 89 | ||
BC048248.1 | - | 5 - 6 | 981 | 6494013 | 89 | |
X57498.1 | - | 5 - 6 | 917 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
